Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIV nef Monoclonal Antibody (17)
GREENER_CHOICE

Catalog No. MA171522 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA171522 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA171522 Supplier Invitrogen™ Supplier No. MA171522
  • $646.00
    $506.65 / Each of 1

    Save $139.35

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

This antibody reacts in ELISA, WB, RIA and IHC with the Nef mac251 strain; it reacts in IF with mac239 and mac1A11 strains. It has been mapped to amino acids 71-80 (-PAEEREKLAY-). This clone may also be referred to as 17 (04-001).

TRUSTED_SUSTAINABILITY

Specifications

Antigen SIV nef
Applications ELISA, Western Blot
Classification Monoclonal
Clone 17
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene Alias Simian Immunodeficiency Virus Nef; simian virus
Host Species Mouse
Immunogen Recombinant SIV-1 Mac251 Nef protein of (purified fromE.coli).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.