Learn More
Bachem H-Arg-Arg-Arg-Arg-Arg-Arg-OH
SDP
Supplier:  Bachem 4028946.0025
Encompass_Preferred
H-Arg-Arg-Arg-Arg-Arg-Arg-OH Trifluoroacetate, 96337-25-6, C36H74N24O7, Mr 955.14, 25mg. Hexaarginine, H(-Arg)6-OH. Six arginines is the minimum number of residues required for obtaining a polycationic peptide which can effectively penetrate cells.
Catalog No. 50-194-6810