Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIX1 Mouse anti-Human, Clone: OTI1H4, lyophilized, TrueMAB™
SDP

Catalog No. 501662607
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-2607 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-2607 Supplier OriGene Supplier No. CF504057
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

The protein encoded by this gene is a homeobox protein that is similar to the Drosophila ′sine oculis′ gene product. This gene is found in a cluster of related genes on chromosome 14 and is thought to be involved in limb development. Defects in this gene are a cause of autosomal domit deafness type 23 (DFNA23) and branchiootic syndrome type 3 (BOS3).

Specifications

Antigen SIX1
Applications Western Blot
Classification Monoclonal
Clone OTI1H4
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene SIX1
Gene Accession No. Q15475
Gene Alias BOS3, DFNA23, TIP39
Gene Symbols SIX1
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-284 of human SIX1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6495
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.