Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIX2 Polyclonal Antibody, Huabio

Catalog No. 502544544
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4544 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4544 Supplier HUABIO RESEARCH INC Supplier No. HA500463
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SIX2 Polyclonal antibody specifically detects SIX2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)

Specifications

Antigen SIX2
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene SIX2
Gene Accession No. Q62232, Q9NPC8
Gene Alias homeobox protein SIX2; Sine oculis homeobox homolog 2; sine oculis-related homeobox 2; sine oculis-related homeobox 2 homolog; SIX homeobox 2; SIX2
Host Species Rabbit
Immunogen Synthetic peptide within mouse six2a aa 100-150/291.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10736, 20472, 366542
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.