Learn More
Selleck Chemical LLC SJ-172550-E0051-5MG
SDP
Supplier:  Selleck Chemical LLC E00515MG
Encompass_Preferred
SJ 172550 is the first MDMX inhibitor that binds reversibly to MDMX and effectively kills retinoblastoma cells in which the expression of MDMX is amplified
Catalog No. 50-234-0766
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.