Learn More
Apexbio Technology LLC SKA 31 40172-65-4 5mg
SDP
Supplier:  Apexbio Technology LLC B74625
Encompass_Preferred
SKA 31 (CAS 40172-65-4) is a small-molecule activator of calcium-activated potassium channels selectively targeting KCa3 1 KCa2 2 KCa2 1 and KCa2 3 subtypes It exhibits EC50 values of 260 nM for KCa3 1 1 9 M for KCa2 2 and 2 9 M for both KCa2 1 and KCa2 3 By facilitating the activation of these channels SKA 31 enhances endothelium-derived hyperpolarizing factor (EDHF) responses This compound is employed to investigate endothelial function and signal transduction with notable utility in studies exploring blood pressure regulation and related vascular mechanisms
Catalog No. 50-000-91995
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.