Learn More
Scott Safety™ SKA-PAK™ Automatic Transfer Supplied Air Respirator
Catalog No. 19121766
Change view
Click to view available options
Duration:
15 min.
3 min.
5 min.
Includes:
Dupont™ Kevlar™ aramid fiber harness, 4500psi carbon 15 min. cylinder, no facepiece, E-Z FLO Vibralert QD regulator, Schrader airline fitting
Nylon harness, 2216psi aluminum bottle, av-2000™ facepiece with Dupont™ Dupont™ Kevlar™ aramid fiber aramid fiber head harness, comfort seal, standard regulator and hansen fitting, vibralert™ tactile alarm system
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Duration Includes
19-121-766 3 min. Nylon harness, 2216psi aluminum bottle, av-2000™ facepiece with Dupont™ Dupont™ Kevlar™ aramid fiber aramid fiber head harness, comfort seal, standard regulator and hansen fitting, vibralert™ tactile alarm system
19-121-861 15 min. Dupont™ Kevlar™ aramid fiber harness, 4500psi carbon 15 min. cylinder, no facepiece, E-Z FLO Vibralert QD regulator, Schrader airline fitting
19-121-768 5 min. Nylon harness, 2216psi aluminum bottle, av-2000™ facepiece with Dupont™ Dupont™ Kevlar™ aramid fiber aramid fiber head harness, comfort seal, standard regulator and hansen fitting, vibralert™ tactile alarm system
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-121-766 Supplier Scott Safety™ Supplier No. SAR422010331101
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Pressure demand supplied air respirator equipped with an automatic, hands-free air transfer capability.
  • Combination Type C positive pressure respirator
  • Automatic, hands-free transfer from external air source to cylinder in the event of primary air source interruption
  • Personal, tactile Vibralert alarm to alert users of transfer to cylinder air and enable prompt escape
  • Onboard cylinder gauge and 90% air remaining lighted alarm for added safety and awareness
  • E-Z Klip cylinder retention system for quick and easy cylinder changes
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance NIOSH
Product Type Automatic Transfer Supplied Air Respirator
Duration 3 min.
For Use With (Application) Confined space, hazardous materials clean-up, chemical, petrochemical, oil and gas, and public utilities.
Includes Nylon harness, 2216psi aluminum bottle, av-2000™ facepiece with Dupont™ Dupont™ Kevlar™ aramid fiber aramid fiber head harness, comfort seal, standard regulator and hansen fitting, vibralert™ tactile alarm system
Model SAR422010331101