Learn More
Scott Safety™ SKA-PAK™ Automatic Transfer Supplied Air Respirator
Description
- Combination Type C positive pressure respirator
- Automatic, hands-free transfer from external air source to cylinder in the event of primary air source interruption
- Personal, tactile Vibralert alarm to alert users of transfer to cylinder air and enable prompt escape
- Onboard cylinder gauge and 90% air remaining lighted alarm for added safety and awareness
- E-Z Klip cylinder retention system for quick and easy cylinder changes
Specifications
Specifications
| Certifications/Compliance | NIOSH |
| Product Type | Automatic Transfer Supplied Air Respirator |
| Duration | 5 min. |
| For Use With (Application) | Confined space, hazardous materials clean-up, chemical, petrochemical, oil and gas, and public utilities. |
| Includes | Nylon harness, 2216psi aluminum bottle, av-2000™ facepiece with Dupont™ Dupont™ Kevlar™ aramid fiber aramid fiber head harness, comfort seal, standard regulator and hansen fitting, vibralert™ tactile alarm system |
| Model | SAR423020332101 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.