Learn More
Scott Safety™ Ska-Pak™ Plus Supplied-Air Respirator
Catalog No. 19122012
Change view
Click to view available options
Pressure:
2216 psig
4500 psig
Includes:
Hansen HK socket, aramid fiber facepiece
Hansen, no facepiece
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pressure Includes
19-122-012 4500 psig Hansen HK socket, aramid fiber facepiece
19-122-014 2216 psig Hansen, no facepiece
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-122-012 Supplier Scott Safety™ Supplier No. SAR324060425301
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Lightweight system with streamlined design for entry into or escape from hazardous, confined space, or Immediately Dangerous to Life and Health (IDLH) situations
  • Features an AV-2000™ full-facepiece with Comfort Seal™ and Kevlar™ head harness for safe, secure fit
  • Allows for interface with communication devices so contact can be maintained in emergencies
  • Comes equipped with the E-Z Klip™ cylinder retention system for quick and easy cylinder changes
  • Cat. No. 19-700-884 includes the Modulair™ Quick Disconnect regulator which allows hook-up to an existing air source such as the Scott Industrial SCBA or E-Z Airline™ supplied air respirators
Order Info
Three styles of harness are available: Kevlar for high heat or spark potential environments; standard nylon; and full-body, with NIOSH-approved integrated respirator design, which holds the cylinders and acts as a fall protection harness. Cylinder options are 5- or 10-minute 3000psig fiberglass cylinders.
Compliance
Meets NFPA airflow performance standards for SCBAs. NIOSH approved.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance NFPA
Product Type Supplied Air Respirator System
Duration 10 min.
For Use With (Application) For entry into or escape from hazardous, confined space, or IDLH situations
For Use With (Equipment) E-Z klip cylinder retention system
Includes Hansen HK socket, aramid fiber facepiece
Model SAR324060425301
Pressure 4500 psig