Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SKI Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520307
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520307 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520307 Supplier Invitrogen™ Supplier No. PA520307
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is human kidney tissue lysate. PA5-20307 can be used with blocking peptide PEP-0401.

TGF-beta is a ubiquitously expressed cytokine that signals through the Smad protein family to regulate numerous cellular processes such as embryonic development and tumorigenesis. This signaling is negatively regulated by Ski, the mammalian homolog of the transforming protein of an avian retrovirus that induces oncogenic transformation of chicken embryo cells, and the related protein SnoN. Ski functions by binding to the Smad proteins and preventing their phosphorylation, thereby inhibiting their ability to bind DNA and activate the transcription of downstream genes. Ski will also bind to the Smad proteins specific to bone morphogenic proteins (BMPs) and block BMP signaling in mammalian cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SKI
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SKI
Gene Accession No. P12755
Gene Alias 2310012I02Rik; 2610001A11Rik; AA062172; AA589460; BC004088; C-ski; Proto-oncogene c-Ski; RGD1565591; SGS; SKI; Ski oncogene; ski oncoprotein; SKI proto-oncogene; ski sarcoma viral oncogene homolog; ski sarcoma viral oncogene homolog (avian); SKV; Sloan-Kettering viral oncogene homolog; v-ski avian sarcoma viral oncogene homolog
Gene Symbols SKI
Host Species Rabbit
Immunogen A 14 amino acid peptide from near the amino terminus of human Ski.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6497
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.