Learn More
PIG™ Skimmer Oil-Only Absorbent Pillow
SDP
Catalog No. 18366624 Shop All New Pig Corporation Products
Change view
Click to view available options
Material:
Polypropylene
Catalog No. Material
18-366-624 Polypropylene
1 options
Catalog No. 18-366-624 Supplier PIG™ Supplier No. PIL405
Only null left
Add to Cart
Add to Cart
High-capacity pillow that absorbs oils and fuels but not water.
  • Pillow has large surface area, high capacity and fast-wicking filler to soak up oils, not water
  • Polypropylene skin resists chemicals and tearing, reduces dust and holds in liquid, even when saturated
  • Ideal for skimming oil off of acid baths or for use in other acidic environments
  • Floats to clean up spills on water confined by booms or socks
  • Skin is UV resistant up to 12 months and meets ANSI and MIL-SPEC standards for static decay
  • Absorbs and retains oils and oil-based liquids, including lubricants, fuels and cleaning agents
  • Bright white color makes pillow easier to see in outdoor environments, draws attention to machine leaks and clearly shows saturation level
  • Can be incinerated after use to reduce waste or for fuels blending
Specifications
Absorbancy 0.5 gal.
Color White
Material Polypropylene
Type Absorbent Pillow
Disposable Single-use
Packaging Quantity 30/Cs.
Dimensions (L x W x H) 12 x 12 x 1 in.
Weight (English) 6 lb.
Weight (Metric) 2.7 kg