Learn More
SKIV2L2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse
Specifications
Specifications
| Antigen | SKIV2L2 |
| Applications | Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Formulation | phosphate, tris citrate with 0.09% sodium azide; pH 7-8 |
| Gene | SKIV2L2 |
| Gene Accession No. | P42285 |
| Gene Alias | ATP-dependent helicase SKIV2L2, ATP-dependent RNA helicase DOB1, ATP-dependent RNA helicase SKIV2L2, Dob1, fSAP118, functional spliceosome-associated protein 118, KIAA0052, Mtr4, superkiller viralicidic activity 2-like 2 (S. cerevisiae), TRAMP-like complex helicase |
| Gene Symbols | SKIV2L2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.