Learn More
Cayman Chemical SKQ1bromIde 25mg
SDP
Supplier:  Cayman Chemical 1989125 MG
Encompass_Preferred
A mitochondrial antioxidant; decreases transmembrane potential and production of ROS; prevents neuronal loss and synaptic damage in rat AD model; slows development of ocular pathologies; prevents acute phenoptosis following rhabdomyolysis, stroke, and myocardial infarction
Catalog No. 50-298-1465
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.