Learn More
Description
- Removable impact-resistant shell with foam lining
- Flanged design for comfort and air flow
- Nylon or hi-vis polyester cap construction with reflective accents
Specifications
Specifications
| Size | Long Brim |
| Color | Lime |
| Quantity | 6/Pk. |
| For Use With (Application) | Baggage Handlers, Manufacturing, Maintenance |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.