Learn More
Ergodyne™ Skullerz™ 8950XL Bump Cap
Catalog No. 19820746 Shop All Ergodyne Products
Change view
Click to view available options
Color:
Navy Blue
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-820-746 -
19-820-744 -
19-820-745 Navy Blue
19-820-747 Navy Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-820-746 Supplier Ergodyne™ Supplier No. 23348
Add to Cart
Add to Cart
Skullerz™ 8950XL Bump Cap provides protection against bumps and bruises.
  • Removable XL impact-resistant ABS plastic shell for larger heads and hairstyles
  • Recommended for hat size 7.5 or above
  • Flanged design for comfort and air flow
  • Nylon cap construction with reflective accents
  • EN 812-compliant