Description
- LED lighting technology built in under brim
- LEDs emit 48 lumens; battery life of approximately 30 continuous hours
- Powered by four CR2032 lithium ion batteries
- Removable impact-resistant shell with foam lining
- Flanged design for comfort and air flow
- Nylon or hi-vis polyester with reflective accents
- EN 812-compliant
Specifications
Specifications
| Color | Lime Green |
| Includes | Under-brim LED lighting technology |
| Quantity | 6/Pk. |
| Size | Adjustable |
| Style | Short Brim |