Learn More
Ergodyne™ Skullerz™ Baldr Safety Glasses/Sunglasses
Catalog No. 19910384 Shop All Ergodyne Products
Change view
Click to view available options
Frame Color:
Black
Kryptek Highlander
Kryptek Typhon
Matte Black
Matte Gray
Lens Tint:
Clear
Clear with Fog-Off™
Copper
Indoor/Outdoor
Indoor/Outdoor with Fog-Off™
Polarized Smoke
Polarized copper
Silver, Mirror
Smoke
Smoke with Fog-Off™
Yellow
42 product options available for selection
Product selection table with 42 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-910-384 Black Polarized Smoke
19-910-379 Black Clear
19-910-380 Black Clear with Fog-Off™
19-910-381 Black Copper
19-910-382 Black Polarized copper
19-910-383 Black Smoke
19-910-385 Black Smoke with Fog-Off™
19-910-386 Black Silver, Mirror
19-910-387 Black Yellow
19-910-388 Black Indoor/Outdoor
19-910-389 Black Indoor/Outdoor with Fog-Off™
19-910-390 Matte Gray Clear
19-910-391 Matte Gray Clear with Fog-Off™
19-910-392 Matte Gray Polarized copper
19-910-393 Matte Gray Smoke
19-910-394 Matte Gray Polarized Smoke
19-910-395 Matte Gray Smoke with Fog-Off™
19-910-396 Matte Gray Yellow
19-910-397 Matte Gray Indoor/Outdoor
19-910-398 Kryptek Highlander Clear
19-910-399 Kryptek Highlander Clear with Fog-Off™
19-910-400 Kryptek Highlander Copper
19-910-401 Kryptek Highlander Polarized copper
19-910-402 Kryptek Highlander Smoke
19-910-403 Kryptek Highlander Polarized Smoke
19-910-404 Kryptek Highlander Smoke with Fog-Off™
19-910-405 Kryptek Highlander Yellow
19-910-406 Matte Black Clear
19-910-407 Matte Black Clear with Fog-Off™
19-910-408 Matte Black Smoke
19-910-409 Matte Black Polarized Smoke
19-910-410 Matte Black Smoke with Fog-Off™
19-910-411 Matte Black Indoor/Outdoor
19-910-412 Matte Black Indoor/Outdoor with Fog-Off™
19-910-413 Kryptek Typhon Clear
19-910-414 Kryptek Typhon Clear with Fog-Off™
19-910-415 Kryptek Typhon Copper
19-910-416 Kryptek Typhon Polarized copper
19-910-417 Kryptek Typhon Smoke
19-910-418 Kryptek Typhon Polarized Smoke
19-910-419 Kryptek Typhon Smoke with Fog-Off™
19-910-420 Kryptek Typhon Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 42 options available.
42 options
This item is not returnable. View return policy
Catalog No. 19-910-384 Supplier Ergodyne™ Supplier No. 57031
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Full-frame protective work safety sunglasses feature tough, scratch-resistant lenses that filter 99.9% of harmful UVA, UVB and UVC rays to keep eyes safe in the sun.
  • Durable frames bend and flex without breaking
  • Non-slip rubber temples keep glasses secure and resist sweat
  • Available with polarized lenses or anti-fogging solution
  • Polarized glasses reduce glare and eye fatigue around water, snow and in other bright environments.
  • Anti-glare glasses are ideal for activities on the water as they block harsh, reflected light from water
  • Fog-Off™ treatment won't wash off or wear off for constant fog-free viewing and are ideal for use with dust masks
  • Certifications and Testing:
    • Tested to ANSI Z87.1+ High-Velocity Impact Protection and optical requirements
    • Tested to MIL-PRF-32432 Ballistic Fragmentation, Class 1, which is the US Military Combat Eye Protection System Performance Specification for eyewear.
    • To pass the military standard, safety glasses must withstand a steel projectile shot at 660 ft / 201m per second which is 4x faster than the Z87+ standard. Lenses must not puncture, crack or shatter. Frames must retain the lenses and not break
  • Ideal for protecting eyes during outdoor work like construction, carpentry/woodworking or landscaping or outdoor hobbies like boating, skiing, fishing, hunting/shooting, and sports.
Specifications
Lens Coating Impact-resistant, Scratch-resistant, UV-treated
Certifications/Compliance ANSI Z87.1+, CSA Z94.3 and MIL-PRF 32432 Ballistic Impact
Lens Tint Polarized Smoke
For Use With (Application) Construction/Trades, Oil and Gas, Mining,Maintenance, Assembly/Fabrication, Manufacturing, Police/Tactical
Frame Color Black
No. per Case 12/Cs.