Learn More
Ergodyne™ Skullerz™ Loki Safety Glasses/Sunglasses
Catalog No. 19910374 Shop All Ergodyne Products
Change view
Click to view available options
Frame Color:
Black
Matte Gray
Lens Tint:
Blue, Mirror
Clear
Clear with Fog-Off™
Copper
Indoor/Outdoor
Indoor/Outdoor with Fog-Off™
Polarized copper
Polarized smoke
Silver, Mirror
Smoke
Smoke with Fog-Off™
Yellow
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-910-374 Matte Gray Smoke
19-910-359 Black Clear
19-910-360 Black Clear with Fog-Off™
19-910-362 Black Polarized copper
19-910-363 Black Smoke
19-910-364 Black Polarized smoke
19-910-365 Black Smoke with Fog-Off™
19-910-368 Black Indoor/Outdoor
19-910-369 Black Indoor/Outdoor with Fog-Off™
19-910-372 Matte Gray Clear with Fog-Off™
19-910-373 Matte Gray Polarized copper
19-910-375 Matte Gray Polarized smoke
19-910-376 Matte Gray Smoke with Fog-Off™
19-910-361 Black Copper
19-910-366 Black Silver, Mirror
19-910-367 Black Yellow
19-910-370 Black Blue, Mirror
19-910-371 Matte Gray Clear
19-910-377 Matte Gray Yellow
19-910-378 Matte Gray Indoor/Outdoor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
This item is not returnable. View return policy
Catalog No. 19-910-374 Supplier Ergodyne™ Supplier No. 56130
This item is not returnable. View return policy
Designed to be worn as regular sunglasses or as safety goggles by removing the temples, attaching the included strap, and adding the removable foam gasket for a dust-beating seal.
  • Protective work safety sunglasses feature tough, scratch-resistant lenses that filter 99.9% of harmful UVA, UVB and UVC rays to keep eyes safe in the sun.
  • Durable frames bend and flex without breaking
  • Non-slip rubber temples keep the glasses secure and resist sweat
  • Available with polarized lenses or with anti-fogging solution
  • Polarized glasses reduce glare and eye fatigue around water, snow and in other bright environments.
  • Anti-glare glasses block the harsh reflected light from water, keeping eyes comfortable around glare.
  • Fog-Off™ treatment won't wash off or wear off and is ideal for use with dust masks.
  • Certifications and Testing:
    • Impact-resistant safety glasses are tested to ANSI Z87.1+ High-Velocity Impact Protection and optical requirements
    • Tested to MIL-PRF-32432 Ballistic Fragmentation, Class 1, which is the US Military Combat Eye Protection System Performance Specification for eyewear. To pass the military standard, safety glasses must withstand a steel projectile shot at 660 ft / 201m per second which is 4x faster than the Z87+ standard. Lenses must not puncture, crack or shatter. Frames must retain the lenses and not break.
  • Ideal for protecting eyes during outdoor work like construction, carpentry/woodworking or landscaping or for outdoor activities like boating, skiing, fishing, hunting/shooting, and sports.
  • Available in a variety of lens options ideally suited for a variety of tasks and light environments.
Specifications
Certifications/Compliance ANSI Z87.1+, CSA Z94.3 and MIL-PRF 32432 Ballistic Impact
Lens Coating Scratch-resistant, UV-treated
Lens Tint Smoke
For Use With (Application) Construction/Trades, Oil and Gas, Mining, Maintenance, Assembly/Fabrication, Manufacturing, Police/Tactical
Frame Color Matte Gray
No. per Case 12/Cs.