Learn More
Ergodyne™ Skullerz™ Odin Safety Glasses/Sunglasses
Catalog No. 19910222 Shop All Ergodyne Products
Change view
Click to view available options
Frame Color:
Black
Kryptek Highlander
Kryptek Typhon
Matte Black
Matte Gray
White
Lens Tint:
Blue, Mirror
Clear
Clear with Fog-Off™
Copper
Indoor/Outdoor
Indoor/Outdoor with Fog-Off™
Polarized Copper
Polarized G15
Polarized Smoke
Silver, Mirror
Smoke
Smoke with Fog-Off™
Yellow
48 product options available for selection
Product selection table with 48 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-910-222 Black Blue, Mirror
19-910-200 Black Clear
19-910-211 Black Clear with Fog-Off™
19-910-213 Black Polarized Copper
19-910-214 Black Smoke
19-910-215 Black Polarized Smoke
19-910-216 Black Smoke with Fog-Off™
19-910-217 Black Silver, Mirror
19-910-218 Black Yellow
19-910-219 Black Polarized G15
19-910-220 Black Indoor/Outdoor
19-910-221 Black Indoor/Outdoor with Fog-Off™
19-910-223 Matte Gray Clear
19-910-224 Matte Gray Clear with Fog-Off™
19-910-225 Matte Gray Polarized Copper
19-910-226 Matte Gray Smoke
19-910-227 Matte Gray Polarized Smoke
19-910-228 Matte Gray Smoke with Fog-Off™
19-910-229 Matte Gray Yellow
19-910-230 Matte Gray Indoor/Outdoor
19-910-231 White Smoke
19-910-232 White Polarized Smoke
19-910-234 Kryptek Highlander Clear
19-910-235 Kryptek Highlander Clear with Fog-Off™
19-910-236 Kryptek Highlander Copper
19-910-237 Kryptek Highlander Polarized Copper
19-910-238 Kryptek Highlander Smoke
19-910-239 Kryptek Highlander Polarized Smoke
19-910-240 Kryptek Highlander Smoke with Fog-Off™
19-910-241 Kryptek Highlander Yellow
19-910-242 Matte Black Clear
19-910-243 Matte Black Clear with Fog-Off™
19-910-244 Matte Black Smoke
19-910-245 Matte Black Polarized Smoke
19-910-246 Matte Black Smoke with Fog-Off™
19-910-247 Matte Black Silver, Mirror
19-910-249 Matte Black Indoor/Outdoor with Fog-Off™
19-910-250 Kryptek Typhon Clear
19-910-251 Kryptek Typhon Clear with Fog-Off™
19-910-252 Kryptek Typhon Copper
19-910-253 Kryptek Typhon Polarized Copper
19-910-254 Kryptek Typhon Smoke
19-910-255 Kryptek Typhon Polarized Smoke
19-910-256 Kryptek Typhon Smoke with Fog-Off™
19-910-257 Kryptek Typhon Yellow
19-910-212 Black Copper
19-910-233 White Silver, Mirror
19-910-248 Matte Black Indoor/Outdoor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 48 options available.
48 options
This item is not returnable. View return policy
Catalog No. 19-910-222 Supplier Ergodyne™ Supplier No. 50092
Add to Cart
Add to Cart
This item is not returnable. View return policy
Full-frame, protective work safety sunglasses feature tough scratch-resistant lenses that filter 99.9% of harmful UVA, UVB and UVC rays to keep eyes safe in the sun.
  • Durable frames bend and flex without breaking
  • Non-slip rubber temples keep the glasses secure and resist sweat
  • Integrated accessory receptors at each temple improve airflow when not in use.
  • For added safety, an elastic retainer strap or foam gasket insert (both sold separately) can be attached to the temples.
  • Available with polarized lenses or with anti-fogging solution
  • Polarized glasses reduce glare and eye fatigue around water, snow and in other bright environments.
  • Anti-glare glasses block the harsh reflected light from water, keeping eyes comfortable around glare.
  • Fog-Off™ treatment won't wash off or wear off and is ideal for use with dust masks.
  • Certifications and Testing:
    • Impact-resistant safety glasses are tested to ANSI Z87.1+ High-Velocity Impact Protection and optical requirements
    • Tested to MIL-PRF-32432 Ballistic Fragmentation, Class 1, which is the US Military Combat Eye Protection System Performance Specification for eyewear. To pass the military standard, safety glasses must withstand a steel projectile shot at 660 ft / 201m per second which is 4x faster than the Z87+ standard. Lenses must not puncture, crack or shatter. Frames must retain the lenses and not break.
  • Ideal for protecting eyes during outdoor work like construction, carpentry/woodworking or landscaping or for outdoor activities like boating, skiing, fishing, hunting/shooting, and sports.
  • Available in a variety of lens options ideally suited for a variety of tasks and light environments.
Specifications
Lens Coating Impact-resistant, Scratch-resistant, UV-treated
Certifications/Compliance ANSI Z87.1+, CSA Z94.3 and MIL-PRF 32432 Ballistic Impact
Lens Tint Blue, Mirror
For Use With (Application) Construction/Trades, Oil and Gas, Mining, Maintenance, Assembly/Fabrication, Manufacturing, Police/Tactical
Frame Color Black
No. per Case 12/Cs.