Learn More
Ergodyne™ Skullerz™ Thor Safety Glasses/Sunglasses
Catalog No. 19910266 Shop All Ergodyne Products
Change view
Click to view available options
Frame Color:
Black
Kryptek Typhon
Matte Gray
Lens Tint:
Blue, Mirror
Clear
Clear with Fog-Off™
Copper
Indoor/Outdoor
Indoor/Outdoor with Fog-Off™
Polarized Copper
Polarized G15
Polarized smoke
Silver, Mirror
Smoke
Smoke with Fog-Off™
Yellow
29 product options available for selection
Product selection table with 29 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-910-266 Black Silver, Mirror
19-910-259 Black Clear
19-910-260 Black Clear with Fog-Off™
19-910-262 Black Polarized Copper
19-910-263 Black Smoke
19-910-264 Black Polarized smoke
19-910-265 Black Smoke with Fog-Off™
19-910-268 Black Polarized G15
19-910-269 Black Indoor/Outdoor
19-910-270 Black Indoor/Outdoor with Fog-Off™
19-910-271 Black Blue, Mirror
19-910-261 Black Copper
19-910-267 Black Yellow
19-910-272 Matte Gray Clear
19-910-273 Matte Gray Clear with Fog-Off™
19-910-274 Matte Gray Polarized Copper
19-910-275 Matte Gray Smoke
19-910-276 Matte Gray Polarized smoke
19-910-277 Matte Gray Smoke with Fog-Off™
19-910-278 Matte Gray Yellow
19-910-279 Matte Gray Indoor/Outdoor
19-910-280 Kryptek Typhon Clear
19-910-281 Kryptek Typhon Clear with Fog-Off™
19-910-282 Kryptek Typhon Copper
19-910-283 Kryptek Typhon Polarized Copper
19-910-284 Kryptek Typhon Smoke
19-910-285 Kryptek Typhon Polarized smoke
19-910-286 Kryptek Typhon Smoke with Fog-Off™
19-910-287 Kryptek Typhon Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 29 options available.
29 options
This item is not returnable. View return policy
Catalog No. 19-910-266 Supplier Ergodyne™ Supplier No. 51042
This item is not returnable. View return policy
Full-frame protective work safety sunglasses feature tough, scratch-resistant lenses filter 99.9% of harmful UVA, UVB and UVC rays to keep your eyes safe in the sun.
  • Durable frames bend and flex without breaking
  • Non-slip rubber nose piece and rubber temples keep the glasses secure and resist sweat
  • Polarized glasses reduce glare and eye fatigue around water, snow and in other bright environments
  • Anti-glare glasses block the harsh reflected light from water, keeping eyes comfortable around glare.
  • Fog-Off™ treatment won't wash off or wear off and is ideal for use with dust masks.
  • Certifications and Testing:
    • Impact-resistant safety glasses are tested to ANSI Z87.1+ High-Velocity Impact Protection and optical requirements
    • Tested to MIL-PRF-32432 Ballistic Fragmentation, Class 1, which is the US Military Combat Eye Protection System Performance Specification for eyewear. To pass the military standard, safety glasses must withstand a steel projectile shot at 660 ft / 201m per second which is 4x faster than the Z87+ standard. Lenses must not puncture, crack or shatter. Frames must retain the lenses and not break.
  • Ideal for protecting eyes during outdoor work like construction, carpentry/woodworking or landscaping or for outdoor activities like boating, skiing, fishing, hunting/shooting, and sports.
  • Available in a variety of lens options ideally suited for a variety of tasks and light environments.
Specifications
Certifications/Compliance ANSI Z87.1+, CSA Z94.3 and MIL-PRF 32432 Ballistic Impact
Lens Tint Silver, Mirror
For Use With (Application) Construction/Trades, Oil and Gas, Mining, Maintenance, Assembly/Fabrication, Manufacturing, Police/Tactical
Frame Color Black
No. per Case 12/Cs.
Frame Material Nylon