Learn More
NOTRAX™ SkyStep™ Rubber Mat
Catalog No. 19086059 Shop All NoTrax Products
Catalog No. 19-086-059 Supplier NOTRAX™ Supplier No. 455S0035BL
Skystep™ is a standalone rubber mat designed for individual work stations
  • Ergonomically engineered
  • Utilizes the power of air
  • Designed to suction to the ground to prevent shifting while trapping air in its chambers
  • Rubber molded, air-pocket underside anti-fatigue mat
  • Made of heavy duty natural rubber compound
  • For general purpose applications
  • Beveled edges on all for sides
Specifications
Color Black
Width (English) 5 ft.
Quantity 1 Ea.
Length (English) 3 ft.