Description
- Ridgeless low profile design
- No rain trough-prevents pooling of liquids
- Soft brow pad absorbs perspiration
Specifications
Specifications
| Color | Red |
| Material | Polyethylene |
| Suspension Type | 4-point ratchet suspension |
| Head Style | Full Brim |