Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLA Class II DR Monoclonal Antibody (2E9/13), FITC
GREENER_CHOICE

Catalog No. PIMA516490 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516490 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516490 Supplier Invitrogen™ Supplier No. MA516490
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Pig SLA Class II DR antibody, clone 289/13 recognizes SLA DR molecules which are expressed on all B cells, antigen presenting cells and on certain subsets of resting and activated T cells.

SLA Class II DR recognizes SLA DR molecules which are expressed on all B cells, antigen presenting cells and on certain subsets of resting and activated T cells. The major histocompatibility complex (MHC) is a cluster of genes that are important in the immune response to infections. In pigs, this is referred to as the swine leukocyte antigen (SLA) region. There are 3 major MHC class II proteins encoded by the SLA which are SLA DP, SLA DQ and SLA DR.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLA Class II DR
Applications Flow Cytometry
Classification Monoclonal
Clone 2E9/13
Concentration 0.1 mg/mL
Conjugate FITC
Formulation PBS with 1% BSA and 0.09% sodium azide
Gene SLA-DRB5
Gene Accession No. 0
Gene Alias DLA class II histocompatibility antigen, DR-1 beta chain-like; DRB1; LA-DRB-c; LA-DRB-d; LOC106504372; MHC class II antigen; MHC class II DR-alpha; MHC class II histocompatibility antigen; MHC class II histocompatibility antigen SLA-DRA; MHC class II histocompatibility antigen SLA-DRB1; MHC class II SLA-DRB1-5; SLA-DR1 beta chain; SLA-DRA; SLA-DRA1; SLA-DRA2; SLA-DR-alpha; SLADRB; SLA-DRB; SLA-DRB*K04G; SLA-DRB*K05C(A); SLA-DRB*K05G; SLA-DRB*K06G; SLA-DRB*K07G; SLA-DRB*K08G; SLA-DRB*K09G; SLA-DRB1; SLA-DR-beta class II light chain
Gene Symbols LOC106504372, SLA-DRA, SLA-DRB1, SLA-DRB2, SLA-DRB3, SLA-DRB4, SLA-DRB5
Host Species Mouse
Immunogen Porcine monocytes.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100135040, 100135041, 100135042, 100135043, 100135047, 100153386, 106504372
Target Species Bovine, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.