Learn More
SLA2 Mouse anti-Human, Clone: OTI1D2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505124

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Adapter protein, which negatively regulates T-cell receptor (TCR) signaling. Inhibits T-cell antigen-receptor induced activation of nuclear factor of activated T-cells. May act by linking signaling proteins such as ZAP70 with CBL, leading to a CBL dependent degradation of signaling proteins.Specifications
SLA2 | |
Monoclonal | |
Unconjugated | |
SLA2 | |
C20orf156, MARS, SLAP-2, SLAP2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
84174 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
OTI1D2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9H6Q3 | |
SLA2 | |
Full length human recombit protein of human SLA2 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.