Learn More
Invitrogen™ SLAMF9 Polyclonal Antibody

Description
A suggested positive control is mouse kidney tissue lysate. PA5-21103 can be used with blocking peptide PEP-1217.
Specifications
Specifications
| Antigen | SLAMF9 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | SLAMF9 |
| Gene Accession No. | Q96A28, Q9D780 |
| Gene Alias | 2310026I04Rik; AI462096; CD2 antigen family, member 10; CD2 family member 10; CD2F10; CD2F-10; CD84 homolog 1; CD84H1; CD84-H1; cluster of differentiation 2 antigen family member 10; cluster of differentiation 84 homolog 1; RP11-48O20.3; SF2001; signaling lymphocytic activation molecule family member 2001; signaling lymphocytic activation molecule family member 9; SLAM family member 9; SLAMF9; UNQ1938/PRO4421 |
| Gene Symbols | SLAMF9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.