Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Slanetz and Bartley Medium (Dehydrated)

Catalog No. OXCM0377B Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXCM0377B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. OXCM0377B Supplier Thermo Scientific™ Supplier No. CM0377B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Detect and enumerate enterococci from water and food samples with this selective medium.

Detect and enumerate enterococci from water and food samples with Thermo Scientific™ Slanetz and Bartley Medium (Dehydrated). Slanetz and Bartley1 originally devised this medium to detect and enumerate enterococci by the technique of membrane filtration, but it has also proved useful as a direct plating medium2, 3. The medium is very selective for enterococci and, when it is incubated at elevated temperatures (44-45°C), all red or maroon colonies may be accepted as presumptive enterococci4, 5.

The detection and enumeration of enterococci is of primary importance for food hygiene monitoring and microbial quality of water and food.

The Environment Agency ‘Microbiology of Drinking Water 2002’6 recommend the use of Slanetz and Bartley medium for the enumeration of enterococci in water supplies, as does ISO in the standard for water quality7. Food samples can be examined for enterococci by the method suggested by the Nordic Committee of Food Analysis3.

  • Easy-to-read: due to the reduction of tetrazolium chloride in the medium the enterococci form dark red/ maroon colonies.
  • Selective: sodium azide in the medium is selective against Gram-negative organisms.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: NMKL.
Description Slanetz and Bartley Medium (Enterococcus Agar, Dehydrated)
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 11.9L medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.