Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC13A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569146
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569146 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569146 Supplier Invitrogen™ Supplier No. PA569146
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 86%; Rabbit: 100%; Rat: 86%.

Mammalian sodium-dicarboxylate cotransporters transport succinate and other Krebs cycle intermediates. They fall into 2 categories based on their substrate affinity: low affinity and high affinity. Both the low- and high-affinity transporters play an important role in the handling of citrate by the kidneys. The protein encoded by this gene represents the high-affinity form. Alternatively spliced transcript variants encoding different isoforms have been found for this gene, although the full-length nature of some of them have not been characterized yet.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC13A3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC13A3
Gene Accession No. Q8WWT9
Gene Alias hNaDC3; hypothetical protein zgc:77173; mNaDC3; Na(+)/dicarboxylate cotransporter 3; Nadc3; NaDC-3; rNaDC3; SDCT2; slc13a3; sodium-dependent high affinity dicarboxylate transporter 3; sodium-dependent high-affinity dicarboxylate transporter 2; sodium-dependent high-affinity dicarboxylate transporter 3; solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 3; solute carrier family 13 member 3; zgc:77173
Gene Symbols SLC13A3
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human SLC13A3.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64849
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.