Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC13A5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560679
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560679 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560679 Supplier Invitrogen™ Supplier No. PA560679
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SQKPKFNFRS QTEEERKTPF YPPPLLDWKV TQEKVPW Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

S13A5 is a tricarboxylate plasma transporter with a preference for citrate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC13A5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC13A5
Gene Accession No. Q86YT5
Gene Alias EIEE25; Indy; mINDY; Na(+)/citrate cotransporter; Na+-coupled citrate transporter protein; NaC2/NaCT; NACT; novel solute carrier family 13 (sodium-dependent dicarboxylate transporter) (Slc13a2 or 3) member; SLC13A5; Sodium-coupled citrate transporter; sodium-dependent citrate transporter; sodium-dependent dicarboxylate transporter; solute carrier family 13 (sodium-dependent citrate transporter), member 5; solute carrier family 13 member 5
Gene Symbols SLC13A5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC13A5. Recombinant protein control fragment (Product #RP-93293).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284111
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.