Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC19A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01674436
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-436 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-436 Supplier Invitrogen™ Supplier No. PA19553
  • $489.00
    $402.30 / Each of 1

    Save $86.70

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

PA1-9553 detects SLC19A1 in human samples. PA1-9553 has been successfully used in Western blot procedures. By Western blot, PA1-9553 detects a 65 kDa protein representing SLC19A1. PA1-9553 immunogen is a purified recombinant SLC19A1 corresponding to the chromosome 21 gene (Human).

Transport of folate compounds into mammalian cells can occur via receptor-mediated or carrier-mediated mechanisms. A functional coordination between these 2 mechanisms has been proposed to be the method of folate uptake in certain cell types. Methotrexate (MTX) is an antifolate chemotherapeutic agent that is actively transported by the carrier-mediated uptake system. RFC1 plays a role in maintaining intracellular concentrations of folate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC19A1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SLC19A1
Gene Accession No. P41440
Gene Alias AI323572; CHMD; FLOT1; folate transporter 1; FOLT; IFC1; IFC-1; Intestinal folate carrier 1; intestinal folate carrier protein; methotrexate carrier 1; methotrexate carrier 2; methotrexate carrier 5; methotrexate carrier 6; MTX1; MTX-1; Placental folate transporter; reduced folate carrier 1; Reduced folate carrier protein; Reduced folate transporter; Reduced folate transporter 1; REFC; RFC; RFC1; RFC-1; RFT-1; SLC19A1; solute carrier family 19 (folate transporter), member 1; solute carrier family 19 (sodium/hydrogen exchanger), member 1; solute carrier family 19 member 1; solute carrier family 19, member 1
Gene Symbols SLC19A1
Host Species Chicken
Immunogen Purified recombinant SLC19A1 corresponding to the chromosome 21 gene (Human).
Purification Method IgY fraction
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6573
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.