Learn More
MBL International Corporation Anti-SLC1A4 (ASCT1) (Human) pAb (Polyclonal Antibody)
SDP
Supplier:  MBL International Corporation BMP028
Encompass_Preferred
Clonality:Polyclonal; Application:FCM|ICC|IHC|WB; Conjugate:Unlabeled; Species Reactivity:Human; Size:50 µl; Target:SLC1A4/ASCT1; Polyclonal antibody of 50 µl targeting SLC1A4/ASCT1 for FCM, ICC, IHC, WB.SLC1A4/ASCT1 Polyclonal Antibody.
Catalog No. 50-453-15
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.