Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ SLC20A1 Recombinant Protein

Catalog No. 89968699 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-968-699 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-968-699 Supplier Abnova™ Supplier No. H00006574P01
Add to Cart
Add to Cart

Used for ELISA, Western Blotting (Recombinant Protein), Antibody Production and Protein Array

Sequence: MATLITSTTAATAASGPLVDYLWMLILGFIIAFVLAFSVGANDVANSFGTAVGSGVVTLKQACILASIFETVGSVLLGAKVSETIRKGLIDVEMYNSTQGLLMAGSVSAMFGSAVWQLVASFLKLPISGTHCIVGATIGFSLVAKGQEGVKWSELIKIVMSWFVSPLLSGIMSGILFFLVRAFILHKADPVPNGLRALPVFYACTVGINLFSIMYTGAPLLGFDKLPLWGTILISVGCAVFCALIVWFFVCPRMKRKIEREIKCSPSESPLMEKKNSLKEDHEETKLSVGDIENKHPVSEVGPATVPLQAVVEERTVSFKLGDLEEAPERERLPSVDLKEETSIDSTVNGAVQLPNGNLVQFSQAVSNQINSSGHYQYHTVHKDSGLYKELLHKLHLAKVGDCMGDSGDKPLRRNNSYTSYTMAICGMPLDSFRAKEGEQKGEEMEKLTWPNADSKKRIRMDSYTSYCNAVSDLHSASEIDMSVKAEMGLGDRKGSNGSLEEWYDQDKPEVSLLFQFLQILTACFGSFAHGGNDVSNAIGPLVALYLVYDTGDVSSKVATPIWLLLYGGVGICVGLWVWGRRVIQTMGKDLTPITPSSGFSIELASALTVVIASNIGLPISTTHCKVGSVVSVGWLRSKKAVDWRLFRNIFMAWFVTVPISGVISAAIMAIFRYVILRM

Specifications

Accession Number AAH19944
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50 mM Tris HCl, 10 mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 6574
Molecular Weight (g/mol) 100.43 kDa
Name SLC20A1 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue
Quantity 2 μg
Source Wheat Germ (in vitro)
Immunogen MATLITSTTAATAASGPLVDYLWMLILGFIIAFVLAFSVGANDVANSFGTAVGSGVVTLKQACILASIFETVGSVLLGAKVSETIRKGLIDVEMYNSTQGLLMAGSVSAMFGSAVWQLVASFLKLPISGTHCIVGATIGFSLVAKGQEGVKWSELIKIVMSWFVSPLLSGIMSGILFFLVRAFILHKADPVPNGLRALPVFYACTVGINLFSIMYTGAPLLGFDKLPLWGTILISVGCAVFCALIVWFFVCPRMKRKIEREIKCSPSESPLMEKKNSLKEDHEETKLSVGDIENKHPVSEVGPATVPLQAVVEERTVSFKLGDLEEAPERERLPSVDLKEETSIDSTVNGAVQLPNGNLVQFSQAVSNQINSSGHYQYHTVHKDSGLYKELLHKLHLAKVGDCMGDSGDKPLRRNNSYTSYTMAICGMPLDSFRAKEGEQKGEEMEKLTWPNADSKKRIRMDSYTSYCNAVSDLHSASEIDMSVKAEMGLGDRKGSNGSLEEWYDQDKPEVSLLFQFLQILTACFGSFAHGGNDVSNAIGPLVALYLVYDTGDVSSKVATPIWLLLYGGVGICVGLWVWGRRVIQTMGKDLTPITPSSGFSIELASALTVVIASNIGLPISTTHCKVGSVVSVGWLRSKKAVDWRLFRNIFMAWFVTVPISGVISAAIMAIFRYVILRM
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias DKFZp686J2397, FLJ41426, GLVR1, Glvr-1, PIT1, PiT-1
Common Name SLC20A1
Gene Symbol SLC20A1
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.