Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC22A10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569255
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569255 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569255 Supplier Invitrogen™ Supplier No. PA569255
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 100%.

SLC22A10 gene ontology annotations related to this gene include transmembrane transporter activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC22A10
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC22A10
Gene Accession No. Q63ZE4
Gene Alias hOAT5; OAT5; Organic anion transporter 5; SLC22A10; solute carrier family 22 (organic anion/cation transporter), member 10; solute carrier family 22 member 10; solute carrier family 22, member 10
Gene Symbols SLC22A10
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human SLC22A10.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387775
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.