Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5 Percent Rhodium on Alumina Powder, Reduced, Dry, 70 microns, BASF Catalyst, Spectrum™ Chemical
SDP

Catalog No. 18607972 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-607-972 50 g
18-607-970 2 g
18-607-971 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-607-972 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SP30150G
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity reduced rhodium catalyst on alumina support for chemical synthesis applications.

  • BASF catalyst containing reduced 5% rhodium compound with particle size of 27 microns

Chemical Identifiers

CAS 7440-16-6
Molecular Formula Rh
Molecular Weight (g/mol) 102.91
MDL Number MFCD00011201 MFCD03458392
InChI Key MHOVAHRLVXNVSD-UHFFFAOYSA-N
IUPAC Name rhodium
SMILES [Rh]

Specifications

Boiling Point 100°C
Physical Form Powder or Liquid
Quantity 50 g
Assay Percent Range 5%
Grade Not Available
Concentration or Composition (by Analyte or Components) 5% Rhodium on Alumina
Packaging Amber Glass Bottle
Appearance Dark Blue Liquid
Decomposition Information No Hazardous Decomposition Products if Stored and Handled as Prescribed or indicated.
Chemical Name or Material 5% Rhodium on Alumina Powder, Reduced, Dry, 70 μ, BASF Catalyst

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.