Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC26A9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562417
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562417 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA562417 Supplier Invitrogen™ Supplier No. PA562417
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MSQPRPRYVV DRAAYSLTLF DDEFEKKDRT YPVGEKLRNA FRC Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.

This gene is one member of a family of sulfate/anion transporter genes. Family members are well conserved in their genomic (number and size of exons) and protein (aa length among species) structures yet have markedly different tissue expression patterns.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC26A9
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC26A9
Gene Accession No. Q7LBE3
Gene Alias Anion transporter/exchanger protein 9; anion transporter/exchanger-9; E030002L01Rik; SLC26A9; SLC26A9 anion transporter/exchanger; solute carrier family 26 (anion exchanger), member 9; solute carrier family 26 member 9; solute carrier family 26, member 9
Gene Symbols SLC26A9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC26A9. Recombinant protein control fragment (Product #RP-99802).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115019
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.