Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC27A6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534544 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534544 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534544 Supplier Invitrogen™ Supplier No. PA534544
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is A-20 cell lysate. PA5-34544 can be used with blocking peptide PEP-1587.

The solute carrier family 27 (fatty acid transporter), member 6 (SLC27A6) is a member of the fatty acid transport protein family (FATP), which are involved in the uptake of long-chain fatty acids. SLC27A6 is principally expressed in the heart on the sarcolemma and colocalizes with CD36, another molecule involved in the uptake of long chain fatty acids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC27A6
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SLC27A6
Gene Accession No. Q9Y2P4
Gene Alias 4732438L20Rik; ACSVL2; acyl-CoA synthetase very long chain family, member 2; FACVL2; FATP1; FATP6; FATP-6; Fatty-acid-coenzyme A ligase, very long-chain 2; long-chain fatty acid transport protein 6; SLC27A6; solute carrier family 27 (fatty acid transporter), member 6; solute carrier family 27 member 6; Very long-chain acyl-CoA synthetase homolog 1; VLCSH1; VLCS-H1
Gene Symbols SLC27A6
Host Species Rabbit
Immunogen A 16 amino acid peptide near the amino terminus of human SLC27A6.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225579, 28965, 291582
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.