Learn More
SLC2A5 Mouse anti-Human, Clone: OTI12F2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500575

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
SLC2A5 (GLUT5) is a cytochalasin B-sensitive carrier. It seems to function primarily as a fructose transporter.Specifications
SLC2A5 | |
Monoclonal | |
Unconjugated | |
SLC2A5 | |
GLUT-5, GLUT5 | |
Mouse | |
Affinity Chromatography | |
RUO | |
6518 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
OTI12F2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P22732 | |
SLC2A5 | |
Full length human recombit protein of human SLC2A5 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.