Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC30A7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113188 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113188 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113188 Supplier Invitrogen™ Supplier No. PA5113188
  • $545.00
    $453.65 / Each of 1

    Save $91.35

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

SLC30A7 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Zinc functions as a cofactor for numerous enzymes, nuclear factors, and hormones and as an intra- and intercellular signal ion. Members of the zinc transporter (ZNT)/SLC30 subfamily of the cation diffusion facilitator family, such as SLC30A7, permit cellular efflux of zinc.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC30A7
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SLC30A7
Gene Accession No. Q8NEW0, Q9JKN1
Gene Alias 1810059J10Rik; 2610034N15Rik; 4833428C12Rik; AI467242; Slc30a7; solute carrier family 30 (zinc transporter), member 7; solute carrier family 30 member 7; Zinc transporter 7; zinc transporter like 2; zinc transporter ZnT-7; ZNT7; znT-7; ZNTL2; Znt-like transporter 2
Gene Symbols SLC30A7
Host Species Rabbit
Immunogen Recombinant Human Zinc transporter 7 protein (162-236AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 148867, 66500
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.