Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC35D1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520554
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520554 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520554 Supplier Invitrogen™ Supplier No. PA520554
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is A-20 cell lysate. PA5-20554 can be used with blocking peptide PEP-0674.

The solute carrier family Slc35 consists of at least 17 proteins that act as nucleotide sugar transporters localized to the Golgi apparatus and endoplasmic reticulum. The role of the ER-resident Slc family member Slc35D1 is to transport both UDP-glucuronic acid and UDP-N-acetylgalactosamine. These molecules can serve as substrates for chondroitin sulfate biosynthesis and mice lacking the Slc35D1 gene developed a lethal form of skeletal dysplasia with severe shortening of limbs and facial structures. Examination of epiphyseal cartilage in these mice revealed a decreased proliferating zone with round chrondrocytes, scarce matrices, and reduced proteoglycan aggregates. Loss of function mutations in human Slc35D1 cause Schneckenbecken dysplasia, a severe skeletal dysplasia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC35D1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SLC35D1
Gene Accession No. A2AKQ0, Q9NTN3
Gene Alias AI834976; C330011J09; KIAA0260; mKIAA0260; SLC35D1; solute carrier family 35 (UDP-GlcA/UDP-GalNAc transporter), member D1; solute carrier family 35 (UDP-glucuronic acid/UDP-N-acetylgalactosamine dual transporter), member D1; solute carrier family 35 member D1; solute carrier family 35, member D1; UDP-galactose transporter-related 7; UDP-galactose transporter-related protein 7; UDP-GlcA/UDP-GalNAc transporter; UDP-glucuronic acid/UDP-N-acetylgalactosamine transporter; UGTREL7
Gene Symbols SLC35D1
Host Species Rabbit
Immunogen A 14 amino acid peptide near the amino terminus of the human Slc35D1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23169, 242585, 298280
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.