Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC36A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542528
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542528 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542528 Supplier Invitrogen™ Supplier No. PA542528
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MSLLGRDYNS ELNSLDNGPQ SPSESSSSIT SENVHPAGEA GLSMMQTLIH Sequence homology: Human: 100%; Pig: 80%.

The function remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC36A3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC36A3
Gene Accession No. Q495N2
Gene Alias PAT3; Proton/amino acid transporter 3; proton-coupled amino acid transporter 3; SLC36A3; solute carrier family 36 (proton/amino acid symporter), member 3; solute carrier family 36 member 3; solute carrier family 36, member 3; TRAMD2; tramdorin 2; tramdorin2; tramdorin-2
Gene Symbols SLC36A3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SLC36A3 (aa 1-50).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285641
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.