Learn More
Invitrogen™ SLC37A1 Polyclonal Antibody
Chicken Polyclonal Antibody
Supplier: Invitrogen™ PA19552
Description
PA1-9552 detects SLC37A1 in human samples. PA1-9552 has been successfully used in Western blot procedures. By Western blot, PA1-9552 detects a 58 kDa protein representing SLC37A1. PA1-9552 immunogen is a purified recombinant SLC37A1 corresponding to the chromosome 21 gene (Human).
SLC37A1, a member of the sugar-phosphate transport family, transports glycerol-3-phosphate (G3P) between cellular compartments for its utilization in several compartment-specific biochemical pathways.
Specifications
SLC37A1 | |
Polyclonal | |
Unconjugated | |
SLC37A1 | |
FLJ22340; G-3-P permease; G-3-P transporter; G3PP; Glucose-6-phosphate exchanger SLC37A1; glycerol-3-phosphate permease; glycerol-3-phosphate transporter; Slc37a1; solute carrier family 37 (glucose-6-phosphate transporter), member 1; solute carrier family 37 (glycerol-3-phosphate transporter), member 1; solute carrier family 37 member 1 | |
Chicken | |
IgY fraction | |
RUO | |
54020 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P57057 | |
SLC37A1 | |
Purified recombinant SLC37A1 corresponding to the chromosome 21 gene (Human). | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgY |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.