Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC44A4 Monoclonal Antibody (3-F2)
GREENER_CHOICE

Catalog No. PIMA542530
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542530 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542530 Supplier Invitrogen™ Supplier No. MA542530
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Positive Control: HepG2 lysates. Subcellular Location: Cell membrane, Membrane.

TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC44A4
Applications Western Blot
Classification Monoclonal
Clone 3-F2
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene SLC44A4
Gene Accession No. Q53GD3
Gene Alias C6orf29; choline transporter-like protein 4; CTL4; hTPPT1; NG22; SLC44A4; Solute carrier family 44 member 4; solute carrier family 44, member 4; testicular tissue protein Li 48; thiamine pyrophosphate transporter; Thiamine pyrophosphate transporter 1; TPPT; TPPT1; UNQ441/PRO874
Gene Symbols SLC44A4
Host Species Mouse
Immunogen Recombinant protein within human SLC44A4 aa 56-229.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80736
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.