Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC47A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554542
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554542 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554542 Supplier Invitrogen™ Supplier No. PA554542
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HANLKVNNVP RSGNSALPQD PLHPGCPENL EGILTNDVGK TGEPQSDQQM RQEEPLPEHP QDGAKLSRKQ Highest antigen sequence identity to the following orthologs: Mouse - 49%, Rat - 51%.

This gene is located within the Smith-Magenis syndrome region on chromosome 17. It encodes a protein of unknown function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC47A1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Slc47a1
Gene Accession No. Q96FL8
Gene Alias 1300013J15Rik; AI036982; FLJ10847; H+/organic cation antiporter variant 1; H+/organic cation antiporter variant 2; hMATE-1; MATE1; MATE-1; MGC64822; mMATE1; mMATE-1; multidrug and toxin extrusion 1; multidrug and toxin extrusion protein 1; RGD1311123; rMATE-1; SLC47A1; solute carrier family 47 (multidrug and toxin extrusion), member 1; solute carrier family 47 member 1; solute carrier family 47, member 1
Gene Symbols Slc47a1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC47A1. Recombinant protein control fragment (Product #RP-93311).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55244
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.