Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC47A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113955
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113955 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113955 Supplier Invitrogen™ Supplier No. PA5113955
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TYSRSECHVD FFRTPEEAHA LSAPTSRLSV KQLVIRRGAA LGAASATLMV For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Dog: 85%; Horse: 79%; Human: 100%.

S47A2 belongs to a family of transporters involved in excretion of toxic electrolytes, both endogenous and exogenous, through urine and bile. This transporter family shares homology with the bacterial MATE (multidrug and toxin extrusion) protein family responsible for drug resistance. This gene is one of two members of the MATE transporter family located near each other on chromosome 17.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC47A2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Slc47a2
Gene Accession No. Q86VL8
Gene Alias 4933429E10Rik; FLJ31196; H(+)/organic cation antiporter kidney-specific; hMATE-2; kidney-specific H(+)/organic cation antiporter; Mate2; MATE-2; MATE2-B; MATE2K; MATE2-K; mMATE-2; multidrug and toxin extrusion protein 2; SLC47A2; solute carrier family 47 (multidrug and toxin extrusion), member 2; solute carrier family 47 member 2; solute carrier family 47, member 2
Gene Symbols Slc47a2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C terminal region of human SLC47A2 (aa 542-591).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146802
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.