Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC4A7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104154 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104154 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104154 Supplier Invitrogen™ Supplier No. PA5104154
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SLC4A7.

SLC4A7 regulates intracellular pH and may also play a role in bicarbonate salvage in secretory epithelia. Additionally, SLC4A7 may also have associated sodium channel activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC4A7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLC4A7
Gene Accession No. Q8BTY2, Q9R1N3, Q9Y6M7
Gene Alias bicarbonate transporter; BT; E430014N10Rik; Electroneutral Na/HCO(3) cotransporter; electroneutral sodium bicarbonate cotransporter 1; Na-bicarbonate cotransporter; Nbc; NBC2; NBC2B; NBC3; NBC-like protein; Nbcn1; NBCn1-G; SBC2; SLC4A6; Slc4a7; Sodium bicarbonate cotransporter 2; sodium bicarbonate cotransporter 2b; Sodium bicarbonate cotransporter 3; sodium bicarbonate cotransporter NBCn1-A; sodium bicarbonate cotransporter NBCn1-B; sodium bicarbonate cotransporter NBCn1-c; sodium bicarbonate cotransporter NBCn1-D; sodium bicarbonate cotransporter NBCn1-H; solute carrier family 4 member 7; solute carrier family 4, sodium bicarbonate cotransporter, member 7
Gene Symbols SLC4A7
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC4A7(Accession Q9Y6M7), corresponding to amino acid residues L377-E427.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117955, 218756, 9497
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.