Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC50A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570596 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570596 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570596 Supplier Invitrogen™ Supplier No. PA570596
  • $584.00
    $478.65 / Each of 1

    Save $105.35

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Dog: 79%; Guinea Pig: 86%; Horse: 91%; Human: 100%; Mouse: 79%; Pig: 93%; Rabbit: 86%; Rat: 86%.

SLC50A1 gene ontology annotations related to this gene include carbohydrate transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC50A1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC50A1
Gene Accession No. Q9BRV3
Gene Alias HsSWEET1; LOW QUALITY PROTEIN: sugar transporter SWEET1; MmSWEET1; probable sugar transporter RAG1AP1; RAG1-activating protein 1; RAG1AP1; recombination activating gene 1 activating protein 1; recombination activating gene 1 gene activation; Rga; RGD1308478; RP11-540D14.5; RZPDo834D038D; SCP; SLC50A1; slv; solute carrier family 50 (sugar efflux transporter), member 1; solute carrier family 50 (sugar transporter), member 1; solute carrier family 50 member 1; Stromal cell protein; sugar transporter SWEET1; SWEET1
Gene Symbols SLC50A1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human SLC50A1 (aa 139-188).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55974
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.