Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC52A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113391
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113391 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113391 Supplier Invitrogen™ Supplier No. PA5113391
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC52A3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

This gene encodes a riboflavin transporter protein that is strongly expressed in the intestine and likely plays a role in intestinal absorption of riboflavin. The protein is predicted to have eleven transmembrane domains and a cell surface localization signal in the C-terminus. Mutations at this locus have been associated with Brown-Vialetto-Van Laere syndrome and Fazio-Londe disease. [provided by RefSeq, Mar 2012].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC52A3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SLC52A3
Gene Accession No. Q9D6X5, Q9NQ40
Gene Alias 2310046K01Rik; bA371L19.1; BVVLS; BVVLS1; C20orf54; ct054; hRFT2; Rft2; RFVT3; RGD1304644; Riboflavin transporter 2; rRFT2; Slc52a3; solute carrier family 52 (riboflavin transporter), member 3; solute carrier family 52 member 3; solute carrier family 52, riboflavin transporter, member 3; solute carrier protein family 52, member 3
Gene Symbols SLC52A3
Host Species Rabbit
Immunogen Recombinant Human Solute carrier family 52, riboflavin transporter, member 3 protein (159-220AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113278, 69698
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.