Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC5A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120634 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120634 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120634 Supplier Invitrogen™ Supplier No. PA5120634
  • $566.00
    $485.65 / Each of 1

    Save $80.35

    14% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: U-87MG, HT-29, 293T, Mouse brain, Rat brain. Immunogen sequence: PPTKEQIRTT TFWSKKNLVV KENCSPKEEP YKMQEKSILR CSENNETINH IIPNGKSEDS IKGLQPEDVN LLVTCREEGN PVASLGHSEA ETPVDAYSNG QAALMGEKER KKETDDGGRY WKFIDWFCGF KSKSLSKRSL RDLMEEEAVC LQMLEETRQV K.

Prevents intracellular accumulation of high concentrations of myo-inositol (an osmolyte) that result in impairment of cellular function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC5A3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene SLC5A3
Gene Accession No. P53794, Q9JKZ2
Gene Alias AA623876; BCW2; BCW2 protein; BF642829; Na(+)/myo-inositol cotransporter; Slc5a3; SMIT; Smit1; SMIT2; sodium myo-inositol transporter 1; sodium/myo-inositol cotransporter; sodium/myo-inositol cotransporter 1; sodium/myo-inositol transporter 1; solute carrier family 5 (inositol transporters), member 3; solute carrier family 5 (sodium/myo-inositol cotransporter), member 3; solute carrier family 5 member 3; solute carrier family 5, member 3; solute carrier family 5, member alpha 3
Gene Symbols SLC5A3
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114507, 53881, 6526
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.