Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC5A6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533045
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533045 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533045 Supplier Invitrogen™ Supplier No. PA533045
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Rat (100%) Monkey, Mouse, Hamster (95%) Marmoset (89%) Bat, Elephant, Panda, Rabbit, Horse (84%).

Transports pantothenate, biotin and lipoate in the presence of sodium. Target protein mediates sodium-dependent transport of pantothenate, biotin, and lipoate; plays a role in intestinal uptake of biotin and in transplacental vitamin transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC5A6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene SLC5A6
Gene Accession No. O70247, Q9Y289
Gene Alias E430023I20; Na(+)-dependent multivitamin transporter; Na+-dependent multivitamin transporter; Slc5a6; SMVT; Sodium-dependent multivitamin transporter; solute carrier family 5 (sodium/multivitamin and iodide cotransporter), member 6; solute carrier family 5 (sodium-dependent vitamin transporter), member 6; solute carrier family 5 member 6; solute carrier family 5, member 6
Gene Symbols SLC5A6
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from N-terminus of human SLC5A6.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170551, 8884
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.