Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC5A7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542485
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542485 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542485 Supplier Invitrogen™ Supplier No. PA542485
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: DDNGIYNQKF PFKTLAMVTS FLTNICISYL AKYLFESGTL PPKLDVFDAV Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

Acetylcholine functions as a neurotransmitter in both the central and peripheral nervous systems of all vertebates, and is the principle neurotransmitter used at the neuromuscular junction. Presynaptic synthesis of acetylcholine requires a steady supply of choline, which is acquired by a plasma membrane high affinity choline transporter (CHT).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC5A7
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Slc5a7
Gene Accession No. Q9GZV3
Gene Alias CHT; CHT1; hCHT; Hemicholinium-3-sensitive choline transporter; high affinity choline transporter 1; high affinity choline transporter 1; LOW QUALITY PROTEIN: high affinity choline transporter 1; high affinity choline transporter; hemicholinium-3-sensitive choline transporter; HMN7A; MGC126299; MGC126300; similar to solute carrier family 5 (choline transporter), member 7; SLC5A7; solute carrier family 5 (choline transporter), member 7; solute carrier family 5 (sodium/choline cotransporter), member 7; solute carrier family 5 member 7; solute carrier family 5, member 7
Gene Symbols Slc5a7
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human SLC5A7.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60482
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.