Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC6A4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585752
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585752 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585752 Supplier Invitrogen™ Supplier No. PA585752
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%), Rhesus Monkey (100%), Bovine (100%), Guinea pig (100%). Positive Control: rat intestine. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

SERT is an integral membrane protein that transports the neurotransmitter serotonin from synaptic spaces into presynaptic neurons. The encoded protein terminates the action of serotonin and recycles it in a sodium-dependent manner. This protein is a target of psychomotor stimulants, such as amphetamines and cocaine, and is a member of the sodium:neurotransmitter symporter family. A repeat length polymorphism in the promoter of this gene has been shown to affect the rate of serotonin uptake and may play a role in sudden infant death syndrome, aggressive behavior in Alzheimer disease patients, and depression-susceptibility in people experiencing emotional trauma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC6A4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 1.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene SLC6A4
Gene Accession No. P31652, Q60857
Gene Alias 5 OCD1; 5HT transporter; 5HTT; 5-HTT; 5-HTTLPR; 5-hydroxytryptamine (serotonin) transporter; AI323329; HGNC:11050; hSERT; HTT; Na+/Cl- dependent serotonin transporter; OCD1; OTTHUMP00000163633; serotonin transporter; serotonin transporter 1; Sert; SERT1; Slc6a4; Sodium-dependent serotonin transporter; Sodium-dependent serotonin transporter (5HT transporter) (5HTT); solute carrier family 6 (neurotransmitter transporter serotonin) member 4 (5-hydroxytryptamine (serotonin) transporter); Solute carrier family 6 (neurotransmitter transporter serotonin) member 4 (5-hydroxytryptamine (serotonin) transporter); solute carrier family 6 (neurotransmitter transporter), member 4; solute carrier family 6 (neurotransmitter transporter, serotonin), member 4; solute carrier family 6 member 4; solute carrier family 6, member 4
Gene Symbols SLC6A4
Host Species Rabbit
Immunogen Synthetic peptide encompassing a sequence within the Extracellular domain of mouse Serotonin transporter. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15567, 25553
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.